Skip to Content
Merck
All Photos(1)

Documents

791300P

Avanti

18:0 PE-DTPA

Avanti Research - A Croda Brand 791300P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H118N9O17P
CAS Number:
Molecular Weight:
1208.55
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (791300P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791300P

shipped in

dry ice

storage temp.

−20°C

General description

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (18:0 PE-DTPA) comprises synthetic phospholipid derivative of ethanolamine.
This chelate is used for preparing MRI contrast agents.

Application

18:0 PE-DTPA (1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (ammonium salt)) has been used to chelate liposome-based cationic adjuvant formulation (CAF01). It has also been used in the liposome preparation for imaging studies

Biochem/physiol Actions

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (18:0 PE-DTPA) or PE-DTPA binding and interaction with the Toll-like receptor-2 regulates immune response . PE-DTPA acts as a lipopolysaccharide antagonist and elicits rescue functionality in sepsis by blocking nuclear factor κ -light-chain-enhancer of activated B cells based transcription.

Packaging

5 mL Amber Glass Screw Cap Vial (791300P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian Mog et al.
Nanotheranostics, 3(4), 342-355 (2019-11-15)
Specific targeting of inflammation remains a challenge in many pathologies, because of the necessary balance between host tolerance and efficacious inflammation resolution. Here, we discovered a short, 4-mer peptide which possesses antagonist properties towards CC chemokine receptor 2 (CCR2), but
Chunghyun Lee et al.
Molecules and cells, 27(2), 251-255 (2009-03-12)
Sepsis is the leading cause of death in critically ill patients. Today, around 60% of all cases of sepsis are caused by Gram-negative bacteria. The cell wall component lipopoly-saccharide (LPS) is the main initiator of the cascade of cellular reactions
Aneesh Thakur et al.
Frontiers in immunology, 9, 2825-2825 (2018-12-18)
Pulmonary tuberculosis (TB), which is caused by Mycobacterium tuberculosis (Mtb), remains a global pandemic, despite the widespread use of the parenteral live attenuated Bacillus Calmette-Guérin (BCG) vaccine during the past decades. Mucosal administration of next generation TB vaccines has great
Jin Young Kang et al.
Immunity, 31(6), 873-884 (2009-11-26)
Toll-like receptor 2 (TLR2) initiates potent immune responses by recognizing diacylated and triacylated lipopeptides. Its ligand specificity is controlled by whether it heterodimerizes with TLR1 or TLR6. We have determined the crystal structures of TLR2-TLR6-diacylated lipopeptide, TLR2-lipoteichoic acid, and TLR2-PE-DTPA
Zahraa S Al-Ahmady et al.
ACS nano, 13(11), 12470-12486 (2019-11-07)
The development of effective therapies for stroke continues to face repeated translational failures. Brain endothelial cells form paracellular and transcellular barriers to many blood-borne therapies, and the development of efficient delivery strategies is highly warranted. Here, in a mouse model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service