Skip to Content
Merck
All Photos(1)

Key Documents

790535P

Avanti

14:0 PE-DTPA

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (ammonium salt), powder

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid; DMPE-DTPA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€139.00

€139.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€139.00

About This Item

Empirical Formula (Hill Notation):
C47H102N9O17P
CAS Number:
Molecular Weight:
1096.34
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€139.00


Estimated to ship on01 June 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (790535P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790535P

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

General description

14:0 PE-DTPA is a lanthanide ion (Ln3+) chelating phospholipid conjugate.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (790535P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication procedures and birefringence measurements for designing magnetically responsive lanthanide ion chelating phospholipid assemblies
Isabettini S, et al.
Journal of Visualized Experiments, e56812-e56812 (2018)
Luke S Evans et al.
Biochimica et biophysica acta. Biomembranes, 1862(9), 183343-183343 (2020-05-18)
Lipidic bicelles have been widely used for the analysis of integral membrane proteins where their spontaneous alignment in an magnetic field has been exploited for oriented sample solid-state NMR studies. Many of their physical properties however make them well suited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service