Skip to Content
Merck
All Photos(1)

Key Documents

890704C

Avanti

18:1 EPC (Cl Salt)

Avanti Research - A Croda Brand 890704C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€176.00
100 MG
€474.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€176.00
100 MG
€474.00

About This Item

Empirical Formula (Hill Notation):
C46H89NO8PCl
CAS Number:
Molecular Weight:
850.63
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€176.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (890704C-25mg)
pkg of 1 × 4 mL (890704C-100mg)
pkg of 2 × 4 mL (890704C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890704C

concentration

10 mg/mL (890704C-25mg)
25 mg/mL (890704C-100mg)
25 mg/mL (890704C-200mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)OCC.[Cl-]

InChI key

LXTQBOAODSPEHG-NATHRRJOSA-M

General description

1,2-dioleoyl-sn-glycero-3-ethylphosphocholine (18:1 EPC) is a cationic alkylated phospholipid.[1]

Application

1,2-dioleoyl-sn-glycero-3-ethylphosphocholine (chloride salt) (18:1 EPC (Cl Salt)) is suitable:
  • for composing lipid shell to prepare lipopolyplex/mRNA vaccine[2]
  • in solvent-assisted preparation of supported lipid bilayers[3]
  • along with 1,2-dioleoyl-sn-glycero-3-phosphatidylethanolamine (DOPE) in mixed liposomes as colloidal vectors for physicochemical study of the DNA compaction[1]
  • to formulate the lipid nanoparticle (LNP1) to enhance intracellular delivery mediated by shock waves[4]

Biochem/physiol Actions

1,2-dioleoyl-sn-glycero-3-ethylphosphocholine (18:1 EPC) serves as a transfection reagent. 18:1 EPC is applicable in liposomes and lipoplexes. It is a potential vector in gene therapy.[1]

Packaging

5 mL Clear Glass Sealed Ampule (890704C-100mg)
5 mL Clear Glass Sealed Ampule (890704C-200mg)
5 mL Clear Glass Sealed Ampule (890704C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene vectors based on DOEPC/DOPE mixed cationic liposomes: a physicochemical study
Munoz U, et al.
Soft Matter, 7(13), 5991-6004 (2011)
Abdul Rahim Ferhan et al.
Nature protocols, 14(7), 2091-2118 (2019-06-09)
The supported lipid bilayer (SLB) platform is a popular cell membrane mimic that is utilized in the chemical, biological, materials science, and medical fields. To date, SLB preparation has proven challenging because of the need for specialized fabrication equipment, domain-specific
Stefano Persano et al.
Biomaterials, 125, 81-89 (2017-02-24)
mRNA-based vaccines have the benefit of triggering robust anti-cancer immunity without the potential danger of genome integration from DNA vaccines or the limitation of antigen selection from peptide vaccines. Yet, a conventional mRNA vaccine comprising of condensed mRNA molecules in
Kabir H Biswas et al.
Langmuir : the ACS journal of surfaces and colloids, 34(4), 1775-1782 (2017-12-28)
The pathway of vesicle adsorption onto a solid support depends on the material composition of the underlying support, and there is significant interest in developing material-independent strategies to modulate the spectrum of vesicle-substrate interactions on a particular surface. Herein, using
Jing Zhang et al.
ACS applied materials & interfaces, 11(11), 10481-10491 (2019-02-23)
Cellular membranes are, in general, impermeable to macromolecules (herein referred to as macrodrugs, e.g., recombinant protein, expression plasmids, or mRNA), which is a major barrier for clinical translation of macrodrug-based therapies. Encapsulation of macromolecules in lipid nanoparticles (LNPs) can protect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service