Skip to Content
Merck
All Photos(2)

Key Documents

700056P

Avanti

lathosterol-d7

Avanti Research - A Croda Brand

Synonym(s):

5α-Cholest-7-en-3β-ol (25,26,26,26,27,27,27-d7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€663.00

€663.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€663.00

About This Item

Empirical Formula (Hill Notation):
C27H39D7O
CAS Number:
Molecular Weight:
393.70
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

€663.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700056P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Lathosterol is a metabolite of cholesterol present in human breast milk.[1] Lathosterol is catabolized by lathosterol 5-desaturase to 7-dehydrocholesterol, which is a precursor of cholesterol.[2] Lathosterol-d7 is a deuterated derivative of lathosterol.

Application

Lathosterol-d7 has been used as an internal standard in gas chromatography-mass spectrometry (GC–MS) for the breast milk sterol quantification[1] and in liquid chromatography high resolution mass spectrometry (LC–HRMS) analysis.[3]

Biochem/physiol Actions

Lathosterol levels are direct indicators of cholesterol levels and aid in decoding metabolism defects. Elevated levels of lathosterol is implicated in cerebrotendinous xanthomatosis (CTX) and lathosterolosis.[4] The inactivation of enzyme lathosterol 5-desaturase (SC5D) leads to accumulation of lathosterol in lathosterolosis pathology.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700056P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrycja A Krakowiak et al.
Human molecular genetics, 12(13), 1631-1641 (2003-06-19)
Lathosterol 5-desaturase catalyzes the conversion of lathosterol to 7-dehydrocholesterol in the next to last step of cholesterol synthesis. Inborn errors of cholesterol synthesis underlie a group of human malformation syndromes including Smith-Lemli-Opitz syndrome, desmosterolosis, CHILD syndrome, CDPX2 and lathosterolosis. We
Monica Gelzo et al.
Lipids in health and disease, 18(1), 235-235 (2020-01-01)
Cerebrotendinous xanthomatosis (CTX) and Lathosterolosis represent two treatable inherited disorders of cholesterol metabolism that are characterized by the accumulation of cholestanol and lathosterol, respectively. The age of the patients suspected of having these disorders is highly variable due to the
Alan H B Wu et al.
Clinica chimica acta; international journal of clinical chemistry, 430, 43-47 (2014-01-08)
The analysis of blood for β-sitosterol and campesterol is the measures of cholesterol absorption while lathosterol is a measure of cholesterol synthesis. The biological variability of β-sitosterol, campesterol, and lathosterol was measured using liquid-chromatography tandem mass spectrometry from a cohort
Andrea Civra et al.
The Journal of steroid biochemistry and molecular biology, 193, 105424-105424 (2019-07-16)
Oxysterols are cholesterol oxidation derivatives. Those containing an additional hydroxyl group on the side chain of the cholesterol molecule result from a physiological enzymatic synthesis and include the majority of oxysterols present in the circulation. Among these, 25-hydroxycholesterol (25OHC) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service