Skip to Content
Merck
All Photos(1)

Key Documents

700118P

Avanti

zymostenol

powder

Synonym(s):

111112

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€377.00

€377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€377.00

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€377.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700118P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700118P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@H]2[C@](CC1)(C3=C([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC2)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-21,23-24,28H,6-17H2,1-5H3/t19-,20+,21+,23-,24+,26+,27-/m1/s1

InChI key

QETLKNDKQOXZRP-XTGBIJOFSA-N

General description

Zymostenol acts as a substrate for Δ8,7-sterol isomerase (EBP).[1] It is a demethylation product of 24,25-dihydrolanosterol (24,25-DHL).[2]

Application

Zymostenol has been used to study its accumulation and its inhibitory actions on Δ8,7-sterol isomerase (EBP).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700118P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diverse Chemical Scaffolds Enhance Oligodendrocyte Formation by Inhibiting CYP51, TM7SF2, or EBP
Allimuthu D, et al.
Cell Chemical Biology (2019)
Insig-mediated degradation of HMG CoA reductase stimulated by lanosterol, an intermediate in the synthesis of cholesterol
Song BL, et al.
Cell Metabolism, 1(3), 179-189 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service