Skip to Content
Merck
All Photos(1)

Documents

700157P

Avanti

Stigmasterol-d5

Avanti Research - A Croda Brand 700157P, powder

Synonym(s):

Stigmasterol-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H43D5O
CAS Number:
Molecular Weight:
417.72
UNSPSC Code:
41141804
NACRES:
NA.25

Assay

>99% (HPLC)

form

powder

packaging

pkg of 1 × 1 mg (700157P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700157P

shipped in

dry ice

storage temp.

−20°C

General description

Stigmasterol-d5 is the deuterated form of the plant sterol, stigmasterol. Stigmasterol is called the “good” sterol. The presence of C24 ethyl group and C22 double bond makes it differ from cholesterol.

Biochem/physiol Actions

Stigmasterol is essential for embryonic development in plants and is also involved in the lipid rafts formation. The presence of an additional ethyl group in stigmasterol promotes membrane cohesion.

Packaging

5 mL Amber Glass Screw Cap Vial (700157P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangfeng Jing et al.
Insect biochemistry and molecular biology, 43(7), 580-587 (2013-04-10)
Insects cannot synthesize sterols de novo, so they typically require a dietary source. Cholesterol is the dominant sterol in most insects, but because plants contain only small amounts of cholesterol, plant-feeding insects generate most of their cholesterol by metabolizing plant
Complex Lipids and Sterols in the Leaves of Eucalyptus macrorhyncha (Myrtaceae) in the Context of Feeding by an Unnamed Gall-Inducing Species of Glycaspis (Synglycaspis)(Hemiptera: Psylloidea: Aphalaridae)
Sharma A, et al.
Annals of the Entomological Society of America, 109(6), 890-898 (2016)
Erick J Dufourc
Plant signaling & behavior, 3(2), 133-134 (2008-02-01)
Membranes of composition approaching those found in "rafts" of plants, fungi and mammals were investigated by means of solidstate (2)H-NMR, using deuterated dipalmitoyl-phosphatidylcholine ((2)H-DPPC) as a reporter. The dynamics of such membranes was determined through measuring of membrane ordering or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service