Skip to Content
Merck
All Photos(1)

Key Documents

610026

Avanti

Extruder Casing-SS

Avanti Research - A Croda Brand

Synonym(s):

Extruder Casing-SS, Stainless Steel Extruder Casing (1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€221.00

€221.00


Check Cart for Availability


Select a Size

Change View
1 EA
€221.00

About This Item

UNSPSC Code:
23151503
NACRES:
NB.51

€221.00


Check Cart for Availability

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

15-25°C

General description

Replacement Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

Extruder Casing-SS has been used to obtain unilamellar vesicles from the acceptor multilamellar vesicles (MLVs) suspension.[1]

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Russel McGuire et al.
The Journal of biological chemistry, 281(6), 3552-3559 (2005-12-13)
Huntingtin-associated protein-1 (HAP1) was initially identified as an interacting partner of huntingtin, the Huntington disease protein. Unlike huntingtin that is ubiquitously expressed throughout the brain and body, HAP1 is enriched in neurons, suggesting that its dysfunction could contribute to Huntington
Milka Doktorova et al.
Nature protocols, 13(9), 2086-2101 (2018-09-08)
Freely suspended liposomes are widely used as model membranes for studying lipid-lipid and protein-lipid interactions. Liposomes prepared by conventional methods have chemically identical bilayer leaflets. By contrast, living cells actively maintain different lipid compositions in the two leaflets of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service