610024
Avanti
Extr. Heating Block
Avanti Research™ - A Croda Brand
Synonym(s):
Extr. Heating Block, Mini-Extruder Holder/Heating Block
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151503
NACRES:
NB.51
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
15-25°C
General description
Accessory Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder
Watch the video to learn more about the Avanti Research™ Mini-Extruder
Biochem/physiol Actions
The Avanti Mini- Extruder allows researchers and scientists to prepare large, unilamellar vesicles by extrusion in an efficient, rapid manner. The optional heating block allows the extrusion of unilamellar vesicles at elevated temperatures, which is critical for the successful production of vesicles from phospholipids with a phase transition temperature (Tm) above room temperature.
Packaging
Original Box from Manufacturer (610024-1Ea)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
also commonly purchased with this product
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service