Skip to Content
Merck
All Photos(1)

Key Documents

610006

Avanti

PC Membranes 0.2um

Avanti Research - A Croda Brand

Synonym(s):

Extruder PC Membrane, Polycarbonate Membranes 0.2μm 19mm (100/pk)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€148.00

€148.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€148.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

€148.00


Check Cart for Availability

Request a Bulk Order

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

PC Membranes are filter membranes with smooth and flat surfaces, with track-etched uniform cylindrical pores.[1]
Polycarbonate Membrane for Avanti′s Mini-Extruder.

Watch the video to learn more about the Avanti Research Mini-Extruder.

Application

PC Membranes 0.2um has been used to extrude large unilamellar vesicles.[2]

Biochem/physiol Actions

PC Membrane exhibits fine mechanical properties, chemical stability, and non-auto fluorescent properties.[3] It is extensively used for the identification of various bacteria by immunofluorescent direct count method.[3]

Components

Whatman® Nuclepore Polyester Drain Discs 10417004

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, lipid membrane incorporation, and ion permeability testing of carbon nanotube porins
Tunuguntla RH, et al.
Nature Protocols, 11(10), 2029-2029 (2016)
Highly sensitive detection of Salmonella typhi using surface aminated polycarbonate membrane enhanced-ELISA
Jain S, et al.
Biosensors And Bioelectronics, 31(1), 37-43 (2012)
Response of MG63 osteoblast-like cells onto polycarbonate membrane surfaces with different micropore sizes
Lee SJ, et al.
Biomaterials, 25(19), 4699-4707 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service