Key Documents
Z675601
Polyethylene carboy with spigot
size 20 L
Select a Size
1 EA
€277.00
€277.00
Check Cart for Availability
Select a Size
Change View
1 EA
€277.00
About This Item
UNSPSC Code:
41121820
NACRES:
NB.35
€277.00
Check Cart for Availability
material
high-density polyethylene (Bottle and spigot)
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F11847-0050
size
20 L
General description
These bottles and carboys have a screw in dispensing spigot that can be replaced with a plug (included) when the vessels are used for long-term storage. Heavy knurling on the cap provides excellent grip for opening and closing. The smooth sides accept adhesive labels and can be marked with most indelible markers.
  • Spigot opens fully in one half-turn
  • Molded of polyethylene, which resists corrosive acids and alkalis
  • Extra heavy walls, yet lighter than glass
Choose from one of the most recent versions:
Certificates of Analysis (COA)