Skip to Content
Merck
All Photos(1)

Key Documents

Z673366

Drilled blocks for Talboys block heater

50 mL conical bottom tube

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€143.00

€143.00


Check Cart for Availability


Select a Size

Change View
1 EA
€143.00

About This Item

UNSPSC Code:
41102404
NACRES:
NB.73

€143.00


Check Cart for Availability

feature

wells 5
Holds 5 x 50 mL tubes

manufacturer/tradename

Troemner 949127

block L × W × H

3.75 in. (9.5 cm) × 3 in. (7.6 cm) × 2 in. (5.1 cm)

well diam. × D

29.0 mm × 47.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

Constructed from a solid anodized aluminum block, the close contact of tubes to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Units are designed as a dry bath/incubator.

Caution

To avoid possible electrical hazard, do not fill well or block with water or other liquids.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service