Z562947
Plas-Labs multi-station Nitrogen dry-box glove box
AC/DC input 120 V AC
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
CE Compliant
description
US plug
manufacturer/tradename
Plas-Labs 850-NBB
H × W × D
28 in. × 60 in. × 38 in., inside dimensions
42 in. × 76 in. × 47 in., outside dimensions
weight
300 lb
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Plas-Labs nitrogen multi-station glove box is a completely enclosed chamber which is ideal for working in a preferred atmosphere. Inert gases like Argon, Nitrogen, and Plasma are easily maintained. It is compact, portable, and lightweight. The design makes is easily moveable from lab to lab.
- Clear, one piece unit with molded base section
- Adjustable SS clamps compensate for wear
- Two pairs of gloves ports for two or more operators working intandem.
- 8 in. O.D. ports with ambidextrous Hypalon® gloves with SS O-rings
- Transparent transfer chamber (12 L × 11 in. i.d.) with adjustable vacuum gauge
- Gas-drying train with DesiSphere desiccating canisters, self-sealing quick-disconnects enable change of drying train without disturbing internal atmosphere
- Two vacuum/pressure pumps - one for transfer chamber, one for drying train to speed purging.
- Fluorescent light system with illuminated controls
- Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere.
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service