Key Documents
Z551864
Sigma-Aldrich
Aldrich® check-valve bubbler with SafetyBarb® hose connectors
single inlet tube, top outlet
Synonym(s):
bubblers, scientific bubbler
Select a Size
1 EA
€258.00
€258.00
Check Cart for Availability
Select a Size
Change View
1 EA
€258.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€258.00
Check Cart for Availability
capacity
15 mL
Related Categories
General description
Check-valve ball prevents oil from being drawn into the system. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)