Key Documents
Z101214
Sigma-Aldrich
Aldrich® in-line bubbler
capacity 5-7 mL
Synonym(s):
bubblers, in-line bubblers, scientific bubblers
Select a Size
1 EA
€135.00
€135.00
Check Cart for Availability
Select a Size
Change View
1 EA
€135.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€135.00
Check Cart for Availability
capacity
5-7 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Dear Sigma Aldrich team, can you please specify the dimensions of this bubbler? Especially the diameter of the inner and outer tubes.
    1 answer
    1. The height of the bubbler is 205 MM, the T-cross has an outer diameter of 8 MM and a inner diameter of 5.2 MM. The side arm has an outer diameter of 8 MM and a inner diameter of 4.8 MM.
      Helpful?