Key Documents
Z530336
Sigma-Aldrich
Aldrich® reaction vessel bubbler
joint: ST/NS 14/20
Synonym(s):
bubblers, scientific bubblers
Select a Size
1 EA
€151.00
€151.00
Check Cart for Availability
Select a Size
Change View
1 EA
€151.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€151.00
Check Cart for Availability
Joint
joint: ST/NS14/20
capacity
5-7 mL
Related Categories
General description
Excellent for closing off a reaction vessel from the atmosphere.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)