Skip to Content
Merck
All Photos(1)

Key Documents

Z265896

Tamer Tabs

good in boiling operations and are highly effective in controlling distillation of liquids

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€19.40

€19.40


Check Cart for Availability


Select a Size

Change View
1 EA
€19.40

About This Item

EC Number:
UNSPSC Code:
41100000
NACRES:
NB.43

€19.40


Check Cart for Availability

mp

2040 °C

General description

Non-reactive, alumina granules are good in boiling operations and are highly effective in controlling distillation of liquids. Avoid applications with strong alkalis and acids, 227g.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service