Skip to Content
Merck
All Photos(1)

Key Documents

1.07912

Supelco

Boiling chips granules

size 1-2 mm

Synonym(s):

Boiling stones

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€52.80

€52.80


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€52.80

About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

€52.80


Estimated to ship on31 May 2025


Request a Bulk Order

size

1-2 mm

Quality Level

storage temp.

no temp limit

General description

~ 1-2 mm

Analysis Note

Sieve analysis (1-2 mm): ≥ 96 %

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service