Key Documents
Z244015
ACE Manifold with threaded stopcocks
positions, 3
Select a Size
1 EA
€972.00
€972.00
Check Cart for Availability
Select a Size
Change View
1 EA
€972.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
€972.00
Check Cart for Availability
feature
positions 3
manufacturer/tradename
Ace Glass 872950
Related Categories
General description
Greaseless vacuum/gas manifold with 0 to 4mm high-vacuum PTFE, three-way stopcocks for takeoffs. One end of each manifold tube has a #7 Ace-Thred for easy cleaning. Ends are supplied with nylon stopper/plugs with FETFE® O-rings. Port connections are 8mm O.D. Vacuum and gas connections are 1/2in. o.d.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)