Skip to Content
Merck

Skip To

Z358193

Nalgene® vacuum filtration system

filter capacity 500 mL, pore size 0.2 μm

Synonym(s):

bottletop filtration system, sterile filtration, Nalgene 566-0020, bottle top strainer, disposable filter unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 case
Check Cart for Availability
€ 310,00

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
Pore size:
0.2 μm
Filter capacity:
500 mL
Packaging:
pack of 12
Material:
polyethersulfone membrane
Sterility:
sterile
Manufacturer/tradename:
Nalgene® 566-0020

€ 310,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyethersulfone membrane

sterility

sterile

feature

cap plug seal, graduations

packaging

pack of 12

manufacturer/tradename

Nalgene® 566-0020

bottle capacity

500 mL

filter capacity

500 mL

funnel capacity

500 mL

membrane diam.

75 mm

pore size

0.2 μm

General description

Increase filtration efficiency by using the Nalgene® Rapid-Flow disposable filter units featuring the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination when filtering cell culture media, serum, additives and buffers. With a graduated upper chamber and Polyethersulfone (PES) membrane this filter unit offer higher aqueous flow rates; lower extractables (no wetting agent); lower protein binding; and greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.

Application

Nalgene™ vacuum filtration system is suitable for the filtration of serum, cell culture medium, tissue culture, buffers and additives.

Features and Benefits

  • Colour coded according to membrane material
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free
  • Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit
  • Graduated Upper chamber

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z370622Z358207Z358185
sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

manufacturer/tradename

Nalgene® 566-0020

manufacturer/tradename

Nalgene® 166-0045

manufacturer/tradename

Nalgene® 567-0020

manufacturer/tradename

Nalgene® 565-0020

pore size

0.2 μm

pore size

0.45 μm

pore size

0.2 μm

pore size

0.2 μm

filter capacity

500 mL

filter capacity

500 mL

filter capacity

1000 mL

filter capacity

150 mL

material

polyethersulfone membrane

material

polyethersulfone membrane

material

polyethersulfone membrane

material

polyethersulfone membrane

packaging

pack of 12

packaging

pack of 12

packaging

pack of 12

packaging

pack of 12


Still not finding the right product?

Explore all of our products under Nalgene® vacuum filtration system




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library