Key Documents
Z183431
Sigma-Aldrich
Powerhold clamp
universal
Select a Size
1 EA
€104.00
€104.00
Check Cart for Availability
Select a Size
Change View
1 EA
€104.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€104.00
Check Cart for Availability
General description
With separate safety stop. Used for mounting rods 3/8 to 5/8 in. diameter. Made from high-strength machined aluminum rod with large knobs for gripping power.
Choose from one of the most recent versions:
Certificates of Analysis (COA)