Z143944
Solid-glass beads
borosilicate, diam. 5 mm
Synonym(s):
borosilicate beads
All Photos(1)
About This Item
UNSPSC Code:
41102422
NACRES:
NB.43
Recommended Products
material
borosilicate
packaging
pack of 1 ea
pack of 4 ea
diam.
5 mm
General description
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Choose from one of the most recent versions:
Certificates of Analysis (COA)