Skip to Content
Merck
All Photos(1)

Key Documents

W206903

Sigma-Aldrich

Isoamyl formate

≥97%, FG

Synonym(s):

Isopentyl formate

Sign Into View Organizational & Contract Pricing

Select a Size

W206903-SAMPLE
€47.40
1 KG
€74.90
9 KG
€425.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W206903-SAMPLE
€47.40
1 KG
€74.90
9 KG
€425.00

About This Item

Linear Formula:
HCOOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2069
Beilstein:
1739893
EC Number:
Council of Europe no.:
500
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.162
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

10 mmHg ( 17.1 °C)

Assay

≥97%

expl. lim.

8 %

refractive index

n20/D 1.397 (lit.)

bp

123-124 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity

SMILES string

[H]C(=O)OCCC(C)C

InChI

1S/C6H12O2/c1-6(2)3-4-8-5-7/h5-6H,3-4H2,1-2H3

InChI key

XKYICAQFSCFURC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service