Skip to Content
Merck
All Photos(1)

Key Documents

W219606

Sigma-Aldrich

Butyl formate

≥97%, FG

Synonym(s):

n-Butyl formate, Butyl menthanoate, Formic acid butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

W219606-SAMPLE
€47.40
1 KG
€118.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W219606-SAMPLE
€47.40
1 KG
€118.00

About This Item

Linear Formula:
HCOO(CH2)3CH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2196
Beilstein:
1742108
EC Number:
Council of Europe no.:
501
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.163
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.389 (lit.)

bp

106-107 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; plum; rum

SMILES string

CCCCOC=O

InChI

1S/C5H10O2/c1-2-3-4-7-5-6/h5H,2-4H2,1H3

InChI key

NMJJFJNHVMGPGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Butyl formate is an aliphatic ester that can be used as a food flavoring agent.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 339-340 (1997)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 339-340 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service