Skip to Content
Merck
All Photos(1)

Key Documents

8.20058

Sigma-Aldrich

Amyl formate

for synthesis

Synonym(s):

Amyl formate, Pentyl formate, Formic acid amyl ester, Formic acid pentyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€101.00

€101.00


Check Cart for Availability


Select a Size

Change View
100 ML
€101.00

About This Item

Linear Formula:
HCOO(CH2)4CH3
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-340-6
NACRES:
NA.22

€101.00


Check Cart for Availability

vapor pressure

70 hPa ( 50 °C)

Quality Level

Assay

≥96.0% (GC)

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-10 % (v/v)

bp

129-131 °C/1013 hPa

mp

-73 °C

transition temp

flash point 26 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCCC)C=O

InChI

1S/C6H12O2/c1-2-3-4-5-8-6-7/h6H,2-5H2,1H3

InChI key

DIQMPQMYFZXDAX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.886 - 0.889
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service