Skip to Content
Merck
All Photos(1)

Key Documents

T78409

Sigma-Aldrich

2,4,4-Trimethyl-1-pentene

2,4,4-Trimethyl-1-pentene

99%

Synonym(s):

α-Diisobutylene, Diisobutylene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€160.00

€160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€160.00

About This Item

Linear Formula:
(CH3)3CCH2C(CH3)=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1098309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€160.00


Check Cart for Availability

Request a Bulk Order

vapor density

>3.8 (vs air)

Quality Level

vapor pressure

83 mmHg ( 37.7 °C)

Assay

99%

autoignition temp.

736 °F

expl. lim.

4.8 %

refractive index

n20/D 1.408 (lit.)

bp

101-102 °C (lit.)

density

0.708 g/mL at 25 °C (lit.)

SMILES string

CC(=C)CC(C)(C)C

InChI

1S/C8H16/c1-7(2)6-8(3,4)5/h1,6H2,2-5H3

InChI key

FXNDIJDIPNCZQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

25.0 °F - closed cup

Flash Point(C)

-3.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diisobutylene.
American Industrial Hygiene Association journal, 43(1), B83-B84 (1982-01-01)
Samuel Barak et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(7), 3451-3460 (2020-02-06)
Soot emissions in combustion are unwanted consequences of burning hydrocarbon fuels. The presence of soot during and following combustion processes is an indication of incomplete combustion and has several negative consequences including the emission of harmful particulates and increased operational

Questions

Reviews

Active Filters

  1. Lusaka Zambia
    • Review 1
    • Vote 1
    5 out of 5 stars.

    This was so good well done

    It is clear and straight forward to understand keep its up

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service