Skip to Content
Merck
All Photos(1)

Key Documents

8.14734

Sigma-Aldrich

2,4,4-Trimethylpentene

(mixture of isomers) for synthesis

Synonym(s):

2,4,4-Trimethylpentene, Diisobutylene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€32.10
1 L
€72.30

€32.10


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€32.10
1 L
€72.30

About This Item

Empirical Formula (Hill Notation):
C8H16
CAS Number:
Molecular Weight:
112.21
MDL number:
UNSPSC Code:
12352002
EC Index Number:
293-261-7
NACRES:
NA.22

€32.10


Estimated to ship on16 May 2025


Request a Bulk Order

vapor pressure

10 hPa ( 20 °C)

Quality Level

Assay

≥90.0% (GC)

form

liquid

autoignition temp.

380 °C (DIN 51794)

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

kinematic viscosity

0.55 cSt

bp

>98 °C/1013 hPa

mp

-106 °C

transition temp

flash point -12 °C

solubility

<0.1 g/L

density

0.72 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.716 - 0.720
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service