Skip to Content
Merck
All Photos(1)

Key Documents

406082

Sigma-Aldrich

2,3,4-Trimethyl-2-pentene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€59.00

€59.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€59.00

About This Item

Linear Formula:
(CH3)2C=C(CH3)CH(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1732093
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€59.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.428 (lit.)

bp

116 °C (lit.)

mp

−113 °C (lit.)

density

0.743 g/mL at 25 °C (lit.)

SMILES string

CC(C)\C(C)=C(/C)C

InChI

1S/C8H16/c1-6(2)8(5)7(3)4/h6H,1-5H3

InChI key

SZFRZEBLZFTODC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,4-Trimethyl-2-pentene is formed during the copolymerization of s-butyl and t-butyl alcohols in the presence of sulfuric acid.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Polymerization of Olefins. II. 1 The Copolymerization of s-Butyl and t-Butyl Alcohols by Sulfuric Acid.
Whitmore FC, et al
Journal of the American Chemical Society, 63(3), 756-757 (1941)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service