Skip to Content
Merck
All Photos(2)

Key Documents

900836

Sigma-Aldrich

4-(1,2,2-Triphenylethenyl)benzaldehyde

Synonym(s):

4-(1,2,2-Triphenylvinyl)benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€165.00

€165.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€165.00

About This Item

Empirical Formula (Hill Notation):
C27H20O
CAS Number:
Molecular Weight:
360.45
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€165.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

Quality Level

form

powder

solubility

organic solvents: soluble (THF, DCM, DMF, DMSO)

storage temp.

2-8°C

SMILES string

[H]C(C(C=C1)=CC=C1C(C2=CC=CC=C2)=C(C3=CC=CC=C3)C4=CC=CC=C4)=O

InChI key

NUUXDUCNYZQXNU-UHFFFAOYSA-N

Application

AIE (aggregation-induced emission)-active building block.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A tetraphenylethene-substituted pyridinium salt with multiple functionalities: synthesis, stimuli-responsive emission, optical waveguide and specific mitochondrion imaging.
Zhao N, et al.
Journal of Material Chemistry C, 1(31), 4640-4646 (2013)
A highly sensitive fluorescent sensor with aggregation-induced emission characteristics for the detection of iodide and mercury ions in aqueous solution.
Zhang RX, et al.
Journal of Material Chemistry C, 4(44), 10479-10485 (2016)
Luminogenic materials constructed from tetraphenylethene building blocks: Synthesis, aggregation-induced emission, two-photon absorption, light refraction, and explosive detection.
Rongrong H, et al.
Journal of Material Chemistry C, 22(1), 232-240 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service