Merck
Skip To
SYNP140100M
Synthware peptide synthesis vessel with fritted disc, T-bore PTFE stopcock and vacuum take-off
100 mL, joint: ST/NS 24/40, porosity: medium, GL 25
Synonym(s):
Synthware P140100M, Solid phase peptide synthesis vessel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 258,00
About This Item
NACRES:
NB.51
UNSPSC Code:
41100000
Joint:
joint: ST/NS24/40
Packaging:
pkg of 1 ea
€ 258,00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P140100M
capacity
100 mL , 100 mL
fitting
GL 25
General description
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas. With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNP140010MSYNP14005MSYNP130100C
manufacturer/tradename
Synthware P140100M
manufacturer/tradename
Synthware P140010M
manufacturer/tradename
Synthware P140050M
manufacturer/tradename
Synthware P130100C
capacity
100 mL , 100 mL
capacity
10 mL , 10 mL
capacity
50 mL , 50 mL
capacity
100 mL , 100 mL
joint
joint: ST/NS24/40
joint
joint: ST/NS24/40
joint
joint: ST/NS24/40
joint
-
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
fitting
GL 25
fitting
GL 14
fitting
GL 25
fitting
GL 25
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
SYNP140100M-1EA04061832197616