Skip to Content
Merck
All Photos(1)

Key Documents

QBD10792

Sigma-Aldrich

m-dPEG®4-thiol

>95% (HPLC)

Synonym(s):

m-PEG4-thiol, mPEG-thiol, mPEG4-SH

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€221.00

€221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€221.00

About This Item

Empirical Formula (Hill Notation):
C9H20O4S
Molecular Weight:
224.32
MDL number:
UNSPSC Code:
12352105
NACRES:
NA.22

€221.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

SCCOCCOCCOCCOC

InChI

1S/C9H20O4S/c1-10-2-3-11-4-5-12-6-7-13-8-9-14/h14H,2-9H2,1H3

InChI key

VXEBAWYHFQRROQ-UHFFFAOYSA-N

Features and Benefits

m-dPEG4-thiol is a monodisperse PEGylation reagent designed to modify gold and silver surfaces or biomolecules that contain free, surface-accessible thiols. The single molecular weight, discrete-length PEG (dPEG) spacer is 14 atoms (15.8 Å) long. One end of the spacer terminates with a methyl group, while the other end terminates with a sulfhydryl. The sulfhydryl group forms dative bonds with gold and disulfides with free thiols under non-reducing conditions.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong Young Choi et al.
ACS nano, 7(3), 2627-2633 (2013-02-14)
In this work, we report a novel polymerase chain reaction (PCR)-free variable-number tandem repeat (VNTR) typing method using a T-shaped gold nanoparticle-DNA monoconjugate, called the "watching-gene assay". The T-shaped DNA probe was synthesized by "click" chemistry and linked with the
Yung-Chieh Chan et al.
ACS omega, 3(2), 1627-1634 (2018-07-20)
The matrix metalloproteinases (MMPs) are well-known mediators that are activated in tumor progression. MMP2 is a kind of gelatinase in extracellular matrix remodeling and cancer metastasis processes. MMP2 secretion increased in many types of cancer diseases, and its abnormal expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service