Skip to Content
Merck
All Photos(1)

Key Documents

736678

Sigma-Aldrich

Tetra(ethylene glycol) dithiol

97%

Synonym(s):

2,2′-[2,2′-Oxybis(ethane-2,1-diyl)bis(oxy)]diethanethiol, Diethylene glycol di(ethanethiol)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€146.00

€146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€146.00

About This Item

Empirical Formula (Hill Notation):
C8H18O3S2
CAS Number:
Molecular Weight:
226.36
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€146.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

refractive index

n20/D 1.502

density

1.100 g/mL at 25 °C

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

SCCOCCOCCOCCS

InChI

1S/C8H18O3S2/c12-7-5-10-3-1-9-2-4-11-6-8-13/h12-13H,1-8H2

InChI key

MCINHQIUGNVTLG-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service