Skip to Content
Merck
All Photos(1)

Key Documents

QBD10850

Sigma-Aldrich

Thiol-dPEG®12-acid

>95% (HPLC)

Synonym(s):

CT(PEG)12, Carboxy-PEG-Thiol, Carboxy-PEG12-Thiol, HS-PEG12-COOH, Thiol-PEG12-acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€367.00

€367.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€367.00

About This Item

Empirical Formula (Hill Notation):
C27H54O14S
Molecular Weight:
634.77
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

€367.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

SCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)O

InChI key

JUDURFKNRMLTLG-UHFFFAOYSA-N

Features and Benefits

Thiol-dPEG12-acid is a single molecular weight, discrete-length polyethylene glycol (dPEG) crosslinking PEGylation reagent. One end of the crosslinker terminates in a sulfhydryl (thiol) group. Propionic acid terminates the opposite end. The thiol end of the molecule forms dative bonds with gold. Moreover, it forms thioether bonds with maleimide and bromoacetate groups and disulfide bonds with other sulfhydryl groups. The propionic acid group reacts with amines to form amide bonds. Alternatively, if it is left unreacted, the propionic acid group provides a negative charge to the conjugated molecule or modified surface. The monodispersed dPEG spacer is 39 atoms (46.8 Å) long.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth R Figueroa et al.
Journal of nanobiotechnology, 14, 24-24 (2016-04-01)
Gold nanoparticles (AuNPs) have shown great promise as scaffolds for gene therapy vectors due to their attractive physiochemical properties which include biocompatibility, ease of functionalization via the nearly covalent gold-sulfur dative bond, and surface plasmon optical properties. Previously, we synthesized
Yung-Chieh Chan et al.
ACS omega, 3(2), 1627-1634 (2018-07-20)
The matrix metalloproteinases (MMPs) are well-known mediators that are activated in tumor progression. MMP2 is a kind of gelatinase in extracellular matrix remodeling and cancer metastasis processes. MMP2 secretion increased in many types of cancer diseases, and its abnormal expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service