Skip to Content
Merck
All Photos(1)

Key Documents

QBD10140

Sigma-Aldrich

Acid-dPEG®25-NHS ester

>95% (HPLC)

Synonym(s):

Acid-PEG-NHS ester, Acid-PEG25-NHS ester, Carboxy-PEG25-NHS ester, NHS-PEG1500-COOH, NHS-PEG25-COOH

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€441.00

€441.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€441.00

About This Item

Empirical Formula (Hill Notation):
C58H109NO31
Molecular Weight:
1316.47
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

€441.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(C(=O)CCC1=O)OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)O

InChI key

XZSKTNJHWDTIJQ-UHFFFAOYSA-N

Features and Benefits

Acid-dPEG25-NHS ester is an amine-reactive crosslinker designed to facilitate intermolecular crosslinking with free amines at each end of the crosslinker through a single molecular weight, discrete polyethylene glycol (dPEG) backbone. The correct use of this product eliminates intramolecular crosslinking. The dPEG backbone is amphiphilic. Consequently, it imparts water solubility to molecules to which it is conjugated. The dPEG spacer is 79 atoms (92.5 Å) long. The dPEG spacer in this product is the monodisperse PEG equivalent of PEG1000. One end of the chain terminates in a propionic acid group. The other end terminates as the N-hydroxysuccinimidyl (NHS) ester of propionic acid. While both ends of the molecule can react with free amines such as the ε-amine group of lysine, only one end of the dPEG spacer is activated as the NHS ester. This construction permits only one end of the spacer to react at a time, thus giving the user greater control over crosslinking molecules using this product.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service