N31400
1-Nonanethiol
95%
Synonym(s):
1-Nonyl mercaptan, Mercaptan C9
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)8SH
CAS Number:
Molecular Weight:
160.32
Beilstein:
1733631
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.455 (lit.)
bp
220 °C (lit.)
density
0.842 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCS
InChI
1S/C9H20S/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3
InChI key
ZVEZMVFBMOOHAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
174.2 °F - closed cup
Flash Point(C)
79 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Steven N Merz et al.
ACS nano, 10(11), 9871-9878 (2016-10-18)
Silver nanoparticles with mixed ligand self-assembled monolayers were synthesized from dodecanethiol and another ligand from a homologous series of alkanethiols (butanethiol, pentanethiol, heptanethiol, octanethiol, nonanethiol, decanethiol, undecanethiol, or dodecanethiol[D25]). These were hypothesized to exhibit ligand phase separation that increases with
Anthony Cirri et al.
Journal of the American Chemical Society, 138(49), 15987-15993 (2016-12-15)
Alkanethiolate protected gold nanoparticles are one of the most widely used systems in modern science and technology, where the emergent electronic properties of the gold core are valued for use in applications such as plasmonic solar cells, photocatalysis, and photothermal
Magdalena Borkowska et al.
Nature nanotechnology, 15(4), 331-341 (2020-03-24)
Lysosomes have become an important target for anticancer therapeutics because lysosomal cell death bypasses the classical caspase-dependent apoptosis pathway, enabling the targeting of apoptosis- and drug-resistant cancers. However, only a few small molecules-mostly repurposed drugs-have been tested so far, and
Hongseung Yeon et al.
Langmuir : the ACS journal of surfaces and colloids, 33(19), 4628-4637 (2017-04-20)
We report an experimental investigation of the influence of molecular-level order (crystallinity) within nonpolar monolayers on hydrophobic interactions. The measurements were performed using gold film-supported monolayers formed from alkanethiols (CH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service