Skip to Content
Merck
All Photos(1)

Key Documents

1.00121

Sigma-Aldrich

Nuclear fast red-aluminum sulfate solution 0.1%

for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€374.00

€374.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€374.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.41

€374.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

concentration

0.1%

technique(s)

microbe id | staining: suitable

density

1.04-1.06 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The ready-to-use Nuclear fast red-aluminum sulfate solution 0.1% for microscopy, is used for counterstaining in histochemical reactions, e.g. in Berlin blue staining for detection of iron or in silver staining of reticulin fibers. Nuclear fast red-aluminum sulfate solution 0.1% can also be used for nuclear staining with Alcian blue. 500 ml of the Nuclear fast red-aluminum sulfate solution 0.1% are sufficient for staining at least 2,500 specimens. The solution has an excellent quality as it is an IVD registered product and is CE certified, thus, it can be used in an accredited laboratory. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

For nuclear staining and for counterstaining in histochemical reactions e.g. Berlin blue

Analysis Note

Suitability for microscopypasses testNucleidark redCytoplasmlight redAcid mucosubstanceslight blue

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service