MT30046252
Mettler Toledo SevenExcellence™ pH/mV conductivity meter S470
S470 kit, meter only, AC/DC input 230 V AC, universal plug set
Select a Size
€3,420.00
Select a Size
About This Item
€3,420.00
Recommended Products
description
Conductivity range 0.001 ?S/cm to 2000 ms/cm, various different units, defined by user (+/- 0.5%)
ISFET pH range 0.000 - 14.000 (+/- 0.05)
Resistivity range 0.01 ... 100.0 MOhm*cm
manufacturer/tradename
Mettler-Toledo 30046252
TDS measuring range
0.001 mg/L (min)
1000 g/L
mV measuring range
-2000-2000, accuracy: ±0.1
salinity measuring range
0-80 ppt
temperature measuring range
-30-130 °C, accuracy: ±0.1
pH range
-2-20
Looking for similar products? Visit Product Comparison Guide
General description
Professional calibration support, incl. automatic calibration with Rondolino sample changer
Flexible due to various conductivity modes, units and temperature correction possibilities
Large 7 inch color display and intuitive menu guidance in 10 languages
Flexible method concept for high reproducibility and security
Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
Integrated interfaces (USB, RS232 and Ethernet) for data exchange
Comprehensive service package including IQ/OQ/PQ
Included in every SevenExcellence:
uPlace™ electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service