Skip to Content
Merck
All Photos(2)

Documents

M52950

Sigma-Aldrich

Methyl isonicotinate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
114618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

207-209 °C (lit.)

mp

8-8.5 °C (lit.)

density

1.161 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccncc1

InChI

1S/C7H7NO2/c1-10-7(9)6-2-4-8-5-3-6/h2-5H,1H3

InChI key

OLXYLDUSSBULGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl isonicotinate can be used in:
  • The preparation of various photosensitizers for applications in dye-sensitized solar cells (DSSCs).
  • Synthesizing luminophore for luminescent metal-organic framework (MOF).
  • Nonaqueous redox flow batteries as pyridine-based anolyte material that undergoes two reversible reductions in solutions with Li-ion electrolytes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evolutionary design of low molecular weight organic anolyte materials for applications in nonaqueous redox flow batteries.
Sevov CS, et al.
Journal of the American Chemical Society, 137(45), 14465-14472 (2015)
Conversion of light to electricity by cis-X2bis (2, 2'-bipyridyl-4, 4'-dicarboxylate) ruthenium (II) charge-transfer sensitizers (X= Cl-, Br-, I-, CN-, and SCN-) on nanocrystalline titanium dioxide electrodes.
Nazeeruddin M K, et al.
Journal of the American Chemical Society, 115(14), 6382-6390 (1993)
Robert W H M van Tol et al.
Entomologia experimentalis et applicata, 168(9), 665-677 (2020-11-06)
'Lure-and-infect' is an insect pest management strategy with high potential but so far there are few examples of its application. Using traps as surrogates for auto-dissemination devices, we tested the attractiveness to naturally occurring thrips (Thysanoptera: Thripidae) of three trap
Luminescent Metal?Organic Frameworks (MOFs) as a Chemopalette: Tuning the Thermochromic Behavior of Dual?Emissive Phosphorescence by Adjusting the Supramolecular Microenvironments.
Zhan SZ, et al.
Chemistry?A European Journal , 19(31), 10217-10225 (2013)
Cyclometalated organoruthenium complexes for application in dye-sensitized solar cells.
Wadman SH, et al.
Organometallics, 29(7), 1569-1579 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service