Skip to Content
Merck
All Photos(2)

Key Documents

151629

Sigma-Aldrich

4-Pyridinemethanol

99%

Synonym(s):

ω-Hydroxy-4-picoline, 4-(Hydroxymethyl)pyridine, 4-Pyridyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€82.10
100 G
€131.00

€82.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€82.10
100 G
€131.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
107850
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€82.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

107-110 °C/1 mmHg (lit.)

mp

52-56 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

OCc1ccncc1

InChI

1S/C6H7NO/c8-5-6-1-3-7-4-2-6/h1-4,8H,5H2

InChI key

PTMBWNZJOQBTBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Pyridinemethanol was used in the preparation of 4-pyridine carboxaldehyde[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F

Flash Point(C)

148 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapour Phase Oxidation of 4-pyridinemethanol to 4-pyridine carboxaldehyde.
Prasad AR and Subrahmanyam M.
Synthetic Communications, 20(21), 3385-3390 (1990)
Lidia Radko et al.
Molecules (Basel, Switzerland), 24(10) (2019-05-24)
In previous papers, we have reported on the high antifungal and significant antibacterial activity against Gram-positive and Gram-negative bacteria of the water-soluble silver(I) complexes of metronidazole and derivatives of pyridine compared to silver nitrate. In the present study, the cytotoxic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service