Skip to Content
Merck
All Photos(1)

Documents

104736

Sigma-Aldrich

Ethyl isonicotinate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
122942
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.501 (lit.)

bp

92 °C/8 mmHg (lit.)

density

1.009 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccncc1

InChI

1S/C8H9NO2/c1-2-11-8(10)7-3-5-9-6-4-7/h3-6H,2H2,1H3

InChI key

MCRPKBUFXAKDKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Martin et al.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989), 49(3), 230-232 (1995-03-01)
The NMDA antagonist CGS 19755 (cis-4-phosphonomethyl-2-piperidinecarboxylic acid) has been prepared by applying Minisci reaction conditions [formamide, hydrogen peroxide, iron(II) sulfate] to ethyl isonicotinate, reduction of the ester with sodium borohydride, alcoholysis of the 2-carboxamide, formation of 4-(diethylphosphonomethyl)-2-pyridinecarboxylate, hydrogenation of the
Gabriella Tamasi et al.
Journal of inorganic biochemistry, 102(10), 1874-1884 (2008-08-08)
The reaction of RuCl(3)(NO).H(2)O with 4-methylpyrimidine (MePYM) and ethylisonicotinate (EINT), in absolute ethanol at 40-55 degrees C afforded crystalline trans-[RuCl(3)(NO)L(2)] complexes. Structural studies via X-ray diffraction, and spectroscopic methods (NMR, IR, UV-visible (UV-Vis)) revealed that the molecular structures have the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service