Skip to Content
Merck
All Photos(1)

Key Documents

J100043

Sigma-Aldrich

Tris(ethylmethylamido)(tert-butylimido)tantalum(V)

electronic grade, ≥95% (CP), ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H33N4Ta
CAS Number:
Molecular Weight:
426.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Assay

≥95% (CP)
≥99.99% trace metals basis

form

liquid

reaction suitability

core: tantalum
reagent type: catalyst

density

1.323 g/mL at 21 °C (lit.)

SMILES string

CCN(C)[Ta](=NC(C)(C)C)(N(C)CC)N(C)CC

InChI

1S/C4H9N.3C3H8N.Ta/c1-4(2,3)5;3*1-3-4-2;/h1-3H3;3*3H2,1-2H3;/q;3*-1;+3

InChI key

PDGHBHKZHSFTHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 73 Tantalum

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

64.0 °F

Flash Point(C)

17.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

High Purity Metalorganic Precursors for CPV Device Fabrication

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service