Skip to Content
Merck
All Photos(1)

Documents

553131

Sigma-Aldrich

Tetrakis(ethylmethylamido)zirconium(IV)

≥99.99% trace metals basis

Synonym(s):

TEMAZ, Tetrakis(ethylmethylamino)zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(NCH3C2H5)4
CAS Number:
Molecular Weight:
323.63
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

liquid

reaction suitability

core: zirconium

bp

81 °C/0.1 mmHg (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

CCN(C)[Zr](N(C)CC)(N(C)CC)N(C)CC

InChI

1S/4C3H8N.Zr/c4*1-3-4-2;/h4*3H2,1-2H3;/q4*-1;+4

InChI key

SRLSISLWUNZOOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrakis(ethylmethylamido)zirconium(IV) can be used as a precursor for atomic layer deposition of zirconium fluoride. These metal fluorides are applied in the field of catalysis, optical films, and protective coatings for Li-ion battery electrodes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZrO2 monolayer as a removable etch stop layer for thermal Al2O3 atomic layer etching using hydrogen fluoride and trimethylaluminum
David R Zywotko, et al.
Chemistry of Materials, 32, 10055-10065 (2020)
Atomic layer deposition of metal fluorides using HF-pyridine as the fluorine precursor
Younghee Lee, et al.
Chemistry of Materials, 28, 2022-2032 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service