Skip to Content
Merck
All Photos(3)

Key Documents

I3502

Sigma-Aldrich

α-Isonitrosopropiophenone

Synonym(s):

1-Phenyl-1,2-propanedione-2-oxime, 2-(Hydroxyimino)propiophenone, NSC 5410

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€45.10
50 G
€152.00

€45.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€45.10
50 G
€152.00

About This Item

Linear Formula:
C6H5COC(=NOH)CH3
CAS Number:
Molecular Weight:
163.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€45.10


Check Cart for Availability

Request a Bulk Order

Quality Level

mp

113-115 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(=N/O)C(=O)c1ccccc1

InChI

1S/C9H9NO2/c1-7(10-12)9(11)8-5-3-2-4-6-8/h2-6,12H,1H3/b10-7+

InChI key

YPINLRNGSGGJJT-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in synthesis of:
  • Cyclometalated iridium complexes for photophysical and electrochemical studies
  • Hydroimidazothiazoles via iodination and cyclization reactions

Reactant involved in:
  • Sonogashira coupling reactions
  • Allylation of aromatic aldehydes using imidazole oside catalysts
  • Beckmann rearrangement of α-oximinoketones

Reagent used in the colorimetric determination of urea

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of N-methylurea: comparison of two colorimetric methods using diacetyl monoxime or alpha-isonitropropiophenone.
W K Paik et al.
Analytical biochemistry, 122(1), 194-198 (1982-05-01)
Luciana Souza-Moreira et al.
Scientific reports, 9(1), 20304-20304 (2020-01-01)
Mesenchymal stromal cells (MSCs) are a potential therapy for many chronic inflammatory diseases due to their regenerative, immunologic and anti-inflammatory properties. The two-way dialogue between MSCs and macrophages is crucial to tissue regeneration and repair. Previous research demonstrated that murine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service