Skip to Content
Merck
All Photos(4)

Key Documents

G9277

Sigma-Aldrich

Guanidine thiocyanate

for molecular biology, ≥99%

Synonym(s):

Guanidinium rhodanide, Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€116.00
250 G
€242.00
500 G
€399.00

€116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€116.00
250 G
€242.00
500 G
€399.00

About This Item

Linear Formula:
NH2C(=NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Beilstein:
3563461
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.31

€116.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic (organic)

Quality Level

grade

for molecular biology

Assay

≥99%

form

powder or crystals

storage condition

(Tightly closed. Dry. )

technique(s)

RNA extraction: suitable

color

colorless to white,

mp

120-122 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless

density

1.29 g/cm3 at 20 °C (68 °F)

cation traces

Ba: ≤5 ppm
Fe: ≤5 ppm
Na: ≤0.5%
heavy metals (as Pb): ≤5 ppm

application(s)

agriculture

foreign activity

DNase, RNase, none detected

SMILES string

SC#N.NC(N)=N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanidine thiocyanate, or guanidinium rhodanide, is a versatile chemical compound widely used in molecular biology and biochemical research. Acting as a chaotropic agent, it disrupts cell and organelle membranes, facilitating cell lysis and organelle isolation. Notably, it serves as a potent protein denaturant, surpassing guanidine hydrochloride in unfolding proteins. This property is valuable for purifying nucleic acids from crude cell lysates, effectively separating nucleic acids from denatured proteins.

In nucleic acid purification, guanidine thiocyanate is effective in inactivating DNases and RNases, preventing nucleic acid degradation and ensuring favorable yields. Its dual denaturing and enzyme-inhibiting properties make it crucial for DNA and RNA studies. Beyond nucleic acid isolation, guanidine thiocyanate is used for cell lysis and virus particle inactivation during RNA and DNA extractions. It not only disrupts membranes but also denatures RNase and DNase enzymes, protecting extracted nucleic acids from enzymatic degradation. The versatility of guanidine thiocyanate extends to molecular biology techniques like PCR, Northern blotting, and Southern blotting. It serves as a vital reagent in these methods, contributing to the successful analysis and amplification of nucleic acids.

Application

Guanidine thiocyanate has been used:
  • for the lysis of colorectal adenocarcinoma cell line prior to RNA isolation[1]
  • as a PCR inhibitor in triplex mitochondrial DNA (mtDNA) qPCR assay[2]
  • for DNA extraction from cerebrospinal fluid (CSF)[3]

Chaotropic agent and strong denaturant; solubilizes cells.

Biochem/physiol Actions

Guanidine thiocyanate is a denaturing agent and is used routinely in RNA isolation. It is used as a storage buffer for whole blood samples.[4] Guanidine thiocyanate inactivates nucleases and is ideal for storing and freezing fecal samples for DNA studies.[5] It is used in combination with phenol−chloroform in RNA extraction.[6]

Features and Benefits

  • Highly versatile surfactant for your cell biology and biochemical research
  • Suitable for RNA extraction in sensitive molecular biology applications
  • Free from Dnase and Rnase
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a universal internal positive control
Kavlick MF
Biotechniques, 65(5), 275-280 (2018)
Direct isolation of poly (A)+ RNA from 4 M guanidine thiocyanate-lysed cell extracts using locked nucleic acid-oligo (T) capture
Jacobsen N, et al.
Nucleic Acids Research, 32(7), e64-e64 (2004)
Comprehensive PCR-based assay for detection and species identification of human herpesviruses
Johnson G, et al.
Journal of Clinical Microbiology, 38(9), 3274-3279 (2000)
Martin Roszkowski et al.
Frontiers in cell and developmental biology, 9, 648274-648274 (2021-05-11)
The extraction of high-quality ribonucleic acid (RNA) from tissues and cells is a key step in many biological assays. Guanidinium thiocyanate-phenol-chloroform (AGPC) is a widely used and efficient method to obtain pure RNA from most tissues and cells. However, it
Abhijeet Singh et al.
Scientific reports, 11(1), 13298-13298 (2021-06-26)
Acetogens play a key role in anaerobic degradation of organic material and in maintaining biogas process efficiency. Profiling this community and its temporal changes can help evaluate process stability and function, especially under disturbance/stress conditions, and avoid complete process failure.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service