Key Documents
DWK247866609
DURAN® desiccator with MOBILEX lid and screw thread closure
thread (DN 250)
Synonym(s):
247866609
Select a Size
1 EACH
€379.00
€379.00
Estimated to ship on14 April 2025
Select a Size
Change View
1 EACH
€379.00
About This Item
UNSPSC Code:
41104510
NACRES:
NB.17
€379.00
Estimated to ship on14 April 2025
material
borosilicate glass 3.3
packaging
case of 1 ea
manufacturer/tradename
DURAN® 247866609
fitting
GL 32 cap
thread (DN 250)
General description
Due to the high wall-thickness of the vessels and the exact machining of the vacuum-tight ground joints on the lid and base, storage under vacuum is possible – even over extremely long periods. Suitable for full vacuum (0 mbar). Spare parts such as lids, bases, stopcocks and caps can be interchanged as required (observe DN). Available with or without porcelain plate. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white print.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)