Key Documents
Z566616
Buchi Combi-Clip
fits 24/40 joint
Select a Size
1 EA
€42.60
€42.60
Check Cart for Availability
Select a Size
Change View
1 EA
€42.60
About This Item
UNSPSC Code:
41104804
NACRES:
NB.43
€42.60
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Büchi 040620
General description
designed to fit the vapor duct for the Rotavapor R-210/215
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–4 of 4 Questions  
  1. Does this clip fit the thread on the RE-111?
    1 answer
    1. This product is a standard Combi-Clip that fits a 24/40 joint, as shown below.
      Helpful?
  2. Ist der Combi-Clip kompatibel mit einem Büchi Rotavapor R-300?
    1 answer
    1. Yes, this combi-clip is compatible with the Büchi Rotavapor R-300.
      Helpful?
  3. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  4. For the Buchi Combi-Clip, Product Z566616, what Buchi model rotary evaporator (rotovap) does it fit?
    1 answer
    1. This Combi-Clip fits only Buchi models R-200/205 and R-210/215. It is usually used with a vapor duct that looks like the one in the attached photo. It will not fit some older Buchi models that have a more narrow duct. The ducts for Buchi models R114/124 (Z403245, short and Z403237, long) already come with a Combi-Clip, so if one of those ducts is ordered, there is no need to order a separate Combi-Clip.
      Helpful?