Skip to Content
Merck
All Photos(2)

Key Documents

DWK218052453

Duran® Gl 45 Laboratory Glass Bottle Protect

plastic-coated glass bottle (PU), with screw cap to drain contents (Without pouring ring), capacity 100 mL

Synonym(s):

218052453

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

plastic-coated glass bottle (PU)

sterility

non-sterile

feature

with screw cap to drain contents (Without pouring ring)

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 218052453

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134 °C.

Application

Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Liquid handling reservoir, Boro 3.3, plastic coated

Features and Benefits

Standardised GL-thread and matching cap system for practicularly tight sealing and simple, plastic coated Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic print; Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service