Skip to Content
Merck
All Photos(2)

Key Documents

D134104

Sigma-Aldrich

2,5-Dimethoxy-2,5-dihydrofuran, mixture of cis and trans

97%

Synonym(s):

2,5-Dihydro-2,5-dimethoxyfuran, DMDF

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€91.70

€91.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€91.70

About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
113225
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€91.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

160-162 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

COC1OC(OC)C=C1

InChI

1S/C6H10O3/c1-7-5-3-4-6(8-2)9-5/h3-6H,1-2H3

InChI key

WXFWXFIWDGJRSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Churchwell et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 86, 1-8 (2015-09-15)
Furan is a food processing contaminant found in many common cooked foods that induces liver toxicity and liver cancer in animal models treated with sufficient doses. The metabolism of furan occurs primarily in the liver where CYP 2E1 produces a
Xu-Dong Wang et al.
Nature communications, 12(1), 1202-1202 (2021-02-24)
Halide perovskite single-crystals have recently been widely highlighted to possess high light harvesting capability and superior charge transport behaviour, which further enable their attractive performance in photovoltaics. However, their application in photoelectrochemical cells has not yet been reported. Here, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service