Skip to Content
Merck
All Photos(1)

Key Documents

8.02961

Sigma-Aldrich

2,5-Dimethoxytetrahydrofuran

(mixture of cis- and trans isomers) for synthesis

Synonym(s):

2,5-Dimethoxytetrahydrofuran

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€34.10
25 ML
€35.60
250 ML
€151.00
50 KG
€7,790.00

€34.10


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€34.10
25 ML
€35.60
250 ML
€151.00
50 KG
€7,790.00

About This Item

Linear Formula:
(OCH3)2C4H6O
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352112
EC Index Number:
211-797-1
NACRES:
NA.22

€34.10


Estimated to ship on17 May 2025


Request a Bulk Order

vapor pressure

5 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

>205 °C

potency

2040 mg/kg LD50, oral (Rat)

expl. lim.

2.0-12.3 % (v/v)

pH

5 (20 °C, 300 g/L in H2O)

bp

145-148 °C/1.013 hPa

mp

-45 °C

transition temp

flash point 35 °C

solubility

300 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(CCC1OC)OC

InChI

1S/C6H12O3/c1-7-5-3-4-6(8-2)9-5/h5-6H,3-4H2,1-2H3

InChI key

GFISDBXSWQMOND-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.022 - 1.024
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service