Skip to Content
Merck
All Photos(2)

Key Documents

H59109

Sigma-Aldrich

3-Hydroxytetrahydrofuran

ReagentPlus®, 98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€25.80

€25.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€25.80

About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€25.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

760 mmHg ( 181 °C)

Quality Level

product line

ReagentPlus®

Assay

98%

refractive index

n20/D 1.45 (lit.)

bp

181 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

OC1CCOC1

InChI

1S/C4H8O2/c5-4-1-2-6-3-4/h4-5H,1-3H2

InChI key

XDPCNPCKDGQBAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service