Skip to Content
Merck
All Photos(1)

Key Documents

D130206

Sigma-Aldrich

2,3-Dimethoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€231.00

About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
908264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€231.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

bp

137 °C/12 mmHg (lit.)

mp

48-52 °C (lit.)

SMILES string

[H]C(=O)c1cccc(OC)c1OC

InChI

1S/C9H10O3/c1-11-8-5-3-4-7(6-10)9(8)12-2/h3-6H,1-2H3

InChI key

JIVGSHFYXPRRSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yadong Li et al.
BMC biochemistry, 12, 30-30 (2011-06-01)
Structural comparison between bacterial CueO and fungal laccases has suggested that a charged residue Glu (E106) in CueO replaces the corresponding residue Phe in fungal laccases at the gate of the tunnel connecting type II copper to the protein surface
Jong H Kim et al.
Annals of clinical microbiology and antimicrobials, 10, 23-23 (2011-06-02)
Disruption of cellular antioxidation systems should be an effective method for control of fungal pathogens. Such disruption can be achieved with redox-active compounds. Natural phenolic compounds can serve as potent redox cyclers that inhibit microbial growth through destabilization of cellular
Olga V Kupriyanova et al.
Drug testing and analysis, 12(8), 1154-1170 (2020-05-18)
N-(2-Methoxybenzyl)-2,5-dimethoxyphenethylamines (NBOMes) are synthetic phenethylamine derivatives emerging on the global drug market and reported to be associated with untoward effects in people who use drugs. Its action involves agonism at serotonin 5-HT2A receptors, affecting cognitive and behavioral processes. However, certain
Jiang Yan et al.
Science advances, 6(45) (2020-11-08)
Phenol is an important commodity chemical in the industry, which is currently produced using fossil feedstocks. Here, we report a strategy to produce phenol from lignin by directly deconstructing Csp2-Csp3 and C-O bonds under mild conditions. It was found that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service